missing translation for 'onlineSavingsMsg'
Learn More

Stachyose tetrahydrate, MedChemExpress

Product Code. 16417527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417527 Supplier Medchem Express Supplier No. HY113529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Stachyose tetrahydrate, a functional oligosaccharide, acts as a prebiotic. Stachyose tetrahydrate can prevent indirectly colon cancer cell growth by promoting the proliferation of probiotics or producing beneficial materials in the intestine.

Stachyose tetrahydrate, a functional oligosaccharide, acts as a prebiotic. Stachyose tetrahydrate can prevent indirectly colon cancer cell growth by promoting the proliferation of probiotics or producing beneficial materials in the intestine.

Specifications

Chemical Name or Material Stachyose tetrahydrate
CAS 10094-58-3
Color White
Purity Grade Notes Research
Molecular Formula C24H50O25
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (338.46 mM; Need ultrasonic) ∣H2O : 100 mg/mL (135.38 mM; Need ultrasonic)
SMILES OC[C@]1(O[C@H](CO)[C@@H](O)[C@@H]1O)O[C@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)CO[C@H]3O[C@@H]([C@H](O)[C@H](O)[C@H]3O)CO[C@H]4O[C@@H]([C@H](O)[C@H](O)[C@H]4O)CO.O.O.O.O
Molecular Weight (g/mol) 738.64
Formula Weight 738.64
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.