missing translation for 'onlineSavingsMsg'
Learn More

Stachydrine hydrochloride, MedChemExpress

Product Code. 16419136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419136 Supplier Medchem Express Supplier No. HYN0738
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Stachydrine hydrochloride is the major active constituent of Herba Leonuri, which is a potential therapy for cardiovascular diseases. Stachydrine can inhibit the NF-κB signal pathway. Anti-hypertrophic activities.

Stachydrine hydrochloride is the major active constituent of Herba Leonuri, which is a potential therapy for cardiovascular diseases. Stachydrine can inhibit the NF-κB signal pathway. Anti-hypertrophic activities.

Specifications

Chemical Name or Material Stachydrine hydrochloride
CAS 4136-37-2
Color White
Purity Grade Notes Research
Molecular Formula C7H14ClNO2
Quantity 100 mg
Solubility Information DMSO : 120 mg/mL (668.00 mM; Need ultrasonic) ∣H2O : 100 mg/mL (556.67 mM; Need ultrasonic)
SMILES OC([C@H](CCC1)[N+]1(C)C)=O.[Cl-]
Molecular Weight (g/mol) 179.64
Formula Weight 179.64
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.