missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565228
This item is not returnable.
View return policy
Description
Immunogen sequence: VCARMIVLNNK FGLRCKNCKT NIHEHCQSYV EMQRCFGKIP PGFHRAYSSP LYSNQQYACV KDLSAANRND PVFET Highest antigen sequence identity to the following orthologs - mouse 91%, rat 95%.
The Src homology 3 (SH3) domain is a highly conserved 60 amino acid protein domain that is organized into a beta-barrel fold consisting of five or six beta strands arranged as two tightly packed anti-parallel beta sheets. This domain is found in proteins that mediate assembly of specific protein complexes and interact with other proteins, specifically recognizing proline-rich regions. STAC3 (SH3 and cysteine rich domain 3) is a 364 amino acid protein containing one phorbol-ester/DAG-type zinc finger and two SH3 (Src homology 3) domains. Existing as two alternatively spliced isoforms, STAC3 maps to human chromosome 12q13.3. Human chromosome 12 encodes over 1,400 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
Specifications
| STAC3 | |
| Polyclonal | |
| Unconjugated | |
| Stac3 | |
| 9830125E18; NAM; SH3 and cysteine rich domain 3; SH3 and cysteine-rich domain-containing protein 3; STAC3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 246329 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MF2 | |
| Stac3 | |
| Recombinant Human STAC3. Recombinant protein control fragment (Product #RP-105960). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction