missing translation for 'onlineSavingsMsg'
Learn More

STAC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18389684 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389684 25 μg 25µL
18396655 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18389684 Supplier Novus Biologicals Supplier No. NBP30966925UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STAC2 Polyclonal specifically detects STAC2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAC2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunoprecipitation
Formulation PBS buffer, 2% sucrose
Gene Alias 24b2/STAC2, MGC129694, SH3 and cysteine rich domain 2,24b2, SH3 and cysteine-rich domain-containing protein 2, Src homology 3 and cysteine-rich domain-containing protein 2
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human STAC2 (NP_945344). Peptide sequence TEMSEKENEPDDAATHSPPGTVSALQETKLQRFKRSLSLKTILRSKSLEN
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342667
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.