missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536512
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.
STAC2 (SH3 and cysteine-rich domain-containing protein 2), also known as 24b2/STAC2, is a 411 amino acid protein that contains one SH3 domain and one phorbol-ester/DAG-type zinc finger. Encoding two alternatively spliced isoforms, the STAC2 gene contains over 15 thousand bases and maps to human chromosome 17q12. Human chromosome 17 comprises over 2.5% of the human genome and encodes over 1, 200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Specifications
| STAC2 | |
| Polyclonal | |
| Unconjugated | |
| STAC2 | |
| 24b2; 24b2/STAC2; AW240854; SH3 and cysteine rich domain 2; SH3 and cysteine-rich domain-containing protein 2; Src homology 3 and cysteine-rich domain-containing protein 2; Stac2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217154, 342667, 363674 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide | |
| Q6ZMT1, Q8R1B0 | |
| STAC2 | |
| Synthetic peptide corresponding to amino acids 212-270 of human STAC2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction