missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11480-1-AP-150UL
This item is not returnable.
View return policy
Description
Probably involved in a neuron-specific signal transduction.Specifications
| STAC | |
| Polyclonal | |
| Unconjugated | |
| STAC | |
| SH3 and cysteine rich domain, STAC, STAC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20840, 363152, 6769 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97306, Q99469 | |
| Stac | |
| STAC Fusion Protein Ag2059 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction