missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557175
This item is not returnable.
View return policy
Description
Immunogen sequence: GSDSPHRTST SDLVEVPEEA NGPGGGYDLR KRSNSVFTYP ENGTDDFRDP AKNINHQGSL SKDPLQMNTY Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.
Promotes expression of the ion channel CACNA1H at the cell membrane, and thereby contributes to the regulation of channel activity. Plays a minor and redundant role in promoting the expression of calcium channel CACNA1S at the cell membrane, and thereby contributes to increased channel activity. Slows down the inactivation rate of the calcium channel CACNA1C. [UniProt].
Specifications
| STAC | |
| Polyclonal | |
| Unconjugated | |
| STAC | |
| SH3 and cysteine rich domain; SH3 and cysteine-rich domain-containing protein; Src homology 3 and cysteine-rich domain-containing protein; src homology three (SH3) and cysteine rich domain; Stac; STAC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6769 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99469 | |
| STAC | |
| Recombinant protein corresponding to Human STAC. Recombinant protein control fragment (Product #RP-96355). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction