missing translation for 'onlineSavingsMsg'
Learn More

Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials

Product Code. 10070654
Change view
Click to view available options
Turbidity:
<0.1, 20, 200, 1000, 4000 NTU
For Use With (Equipment):
Hach 2100N Turbidimeter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Turbidity For Use With (Equipment) Unit Size
10070654 <0.1, 20, 200, 1000, 4000 NTU Hach 2100N Turbidimeter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10070654 Supplier Hach™ Supplier No. 2662105
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Developed for use in any turbidimeter. Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials are true Formazin dilutions in individual sealed vials of <0.1, 20, 200, 1000, 4000 NTU and 7500 NTU (2662105 only) levels.

  • Precise, Pre-mixed Formazin Solutions: Provides accurate turbidimeter calibrations
  • Based on Formazin Primary Standards: Contains the same light scattering polymer and performs equivalently
  • Enhanced Stability Saves Time and Labor: Repeatable results without preparing standards in the lab
  • Confidence in Results: Based on decades of experience in the drinking water industry
  • Any Turbidity Meter: Can be used with 2100N/2100AN or 2100N/2100AN IS Turbidimeter
TRUSTED_SUSTAINABILITY

Specifications

Type Turbidity Standard Kit
For Use With (Equipment) Hach 2100N Turbidimeter
Includes Premixed, sealed vials
For Use With (Application) Calibration of turbidimeters
Turbidity <0.1, 20, 200, 1000, 4000 NTU
compliance-icons
Product Identifier
  • StablCal Formazin Turbidity Standard
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P363-Wash contaminated clothing before reuse.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: methenamine, hexamethylenetetramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.