missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Stablcal™ Stabilized Formazine Turbidity Standard 1 NTU

Product Code. 10622123
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10622123 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10622123 Supplier Hach Lange™ Supplier No. 2659842
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hach Lange Stablcal Stabilized Formazine Turbidity Standard 1 NTU, Stablcal Stabilized Formazine Turbidity Standard 1.0 NTU, Volume: 100mL

TRUSTED_SUSTAINABILITY

Specifications

Type Stablcal Stabilized Formazine Turbidity Standard
Turbidity 1.0 NTU
Volume (Metric) 100 mL
compliance-icons
Product Identifier
  • StablCal Standard, 1.0 NTU
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Formaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.