missing translation for 'onlineSavingsMsg'
Learn More

Hach Pacific Pty Ltd Stablcal Ampule Calibration Kit, Formazin, for 2100Q/QIS portable turbidimeter

Product Code. 16215530
Change view
Click to view available options
For Use With (Equipment):
Portable Turbidimeter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16215530 Portable Turbidimeter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16215530 Supplier Hach Pacific Pty Ltd Supplier No. 2971205
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calibration Standard kit for 2100Q/QIS portable turbidimeter.

Calibration Standard kit for 2100Q/QIS portable turbidimeter. Ampule kit contains Formazin standards in concentrations: 10, 20, 100, 800 NTU. Precise, Pre-mixed Formazin Solutions Based on Formazin Primary Standards Enhanced Stability Saves Time and Labor Confidence in results

Precise, Pre-mixed Formazin Solutions
With proprietary manufacturing technology, Hach prepares Stablcal Standards in precise concentrations at a wide variety of concentrations - providing accurate turbidimeter calibrations.

Based on Formazin Primary Standards
Stablcal Stabilized Formazin Standards contain the same light scattering polymer as traditional Formazin primary turbidity standards - and perform equivalently.

Enhanced Stability Saves Time and Labor
While Formazin primary standards can be prepared directly in the lab, the process is labor intensive and time consuming - and the diluted standards are not stable. Stablcal is a stabilized version of Formazin - providing repeatable results, without the time and effort required for lab-prepared standards.

Confidence in results
Hach was the first to apply Formazin as a turbidity standard in the drinking water industry, more than 40 years ago. Stablcal standards are based on decades of experience in turbidity measurement.

Specifications
Concentration: 20, 100, 800, 10 NTU (for verification)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Portable Turbidimeter
compliance-icons
Product Identifier
  • STABLCAL Turbidity Standards Calibration Kit
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methenamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.