missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stabilizing Fixative, BD
Click to view available options
Quantity:
90 mL
Unit Size:
Each
Specifications
Specifications
| Chemical Name or Material | Stabilizing Fixative |
| For Use With (Application) | Preserving immunofluorescence staining of human peripheral blood cells with monoclonal antibodies prior to flow cytometric analysis |
| Quantity | 90 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction