missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stabilin 2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547522
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human Stabilin-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Stabilin 2 Polyclonal antibody specifically detects Stabilin 2 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
Stabilin 2 | |
Polyclonal | |
Unconjugated | |
STAB2 | |
190 kDa form stabilin-2; 190 kDa hyaluronan receptor for endocytosis; CD44-like precursor FELL; DKFZP434E0321; FAS1 EGF-like and X-link domain containing adhesion molecule-2; FAS1 EGF-like and X-link domain-containing adhesion molecule 2; fasciclin egf-like, laminin-type egf-like, and link domain-containing scavenger receptor-2; fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 2; FEEL2; FEEL-2; FELE-2; FELL; FELL2; FELL-2; FEX2; Hare; HAREFELL2; hepatic hyaluronan clearance receptor; hyaluronan receptor for endocytosis; hyaluronic acid receptor for endocytosis; MFEEL-2; SCARH1; scavenger receptor FEEL-2; Short form stabilin-2; stab2; STAB-2; stab2 {ECO:0000250; stabilin 2; Stabilin2; stabilin-2; UniProtKB:Q8WWQ8} | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
55576 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8WWQ8 | |
STAB2 | |
E. coli-derived recombinant human Stabilin-2 Gly2198-Val2295 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |