missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Stabilin-1 Antibody (bexmarilimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28672-50ug
This item is not returnable.
View return policy
Description
Stabilin-1 Monoclonal antibody specifically detects Stabilin-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Stabilin-1 | |
| Monoclonal | |
| Unconjugated | |
| Q9NY15 | |
| Human | |
| Protein A purified | |
| RUO | |
| 23166 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| bexmarilimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CLEVER-1, fasciclin egf-like, laminin-type egf-like, and link domain-containing scavengerreceptor-1, Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavengerreceptor 1, FEEL1, FEEL-1MS-1 antigen, FELE-1, FEX1, KIAA0246common lymphatic endothelial and vascular endothelial receptor-1, STAB-1, stabilin 1, stabilin-1 | |
| STAB1 | |
| 50 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction