missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST8SIA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30493-25ul
This item is not returnable.
View return policy
Description
ST8SIA5 Polyclonal specifically detects ST8SIA5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ST8SIA5 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| O15466 | |
| ST8SIA5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RYFEFYEGPFEYNSTRCLELRHEILEVKVLSMVKQSELFDRWKSLQMCKWAMNISEANQFKSTLS | |
| 25ul | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| alpha-2,8-sialyltransferase 8E, EC 2.4.99.-, MGC119670, Sialyltransferase 8E, sialyltransferase 8E (alpha-2, 8-polysialytransferase), Sialytransferase St8Sia V, SIAT8-E, SIAT8EMGC119671, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5, ST8SiaV | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 29906 | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering