missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST8SIA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566866
This item is not returnable.
View return policy
Description
Immunogen sequence: WPFGFDPNTRE DLPYHYYDKK GTKFTTKWQE SHQLPAEFQL LYRMHGEGLT KLTLSHCA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
ST8SIA3 belongs to a family of sialyltransferases that form sialyl-alpha-2,8-sialyl-R linkages at the nonreducing termini of glycoconjugates.
Specifications
| ST8SIA3 | |
| Polyclonal | |
| Unconjugated | |
| St8sia3 | |
| AI847333; Alpha-2,8-sialyltransferase 8C; alpha-2,8-sialyltransferase III; GT3-synthase; sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase; sialyltransferase 8 (alpha-2, 8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 2,8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 28-sialyltransferase) C; sialyltransferase 8 C; sialyltransferase 8C; sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase); sialyltransferase St8Sia III; sialytransferase St8Sia III; SIAT8C; SIAT8-C; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 3; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3; St8sia3; ST8SiaIII | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51046 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43173 | |
| St8sia3 | |
| Recombinant Human ST8SIA3. Recombinant protein control fragment (Product #RP-107518). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction