missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST8SIA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | ST8SIA3 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261414
|
Novus Biologicals
NBP2-56457 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670309
|
Novus Biologicals
NBP2-56457-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ST8SIA3 Polyclonal specifically detects ST8SIA3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ST8SIA3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Alpha-2,8-sialyltransferase 8C, Alpha-2,8-sialyltransferase III, EC 2.4.99.-, sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase, Sialyltransferase 8C, sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase), Sialytransferase St8Sia III, SIAT8-C, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3SIAT8C, ST8SiaIII | |
| ST8SIA3 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 51046 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:WPFGFDPNTREDLPYHYYDKKGTKFTTKWQESHQLPAEFQLLYRMHGEGLTKLTLSHCA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title