missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6GALNAC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16442-1-AP-20UL
This item is not returnable.
View return policy
Description
ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions. ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions (Tsuchida et al., 2003 ).Specifications
| ST6GALNAC5 | |
| Polyclonal | |
| Unconjugated | |
| ST6GALNAC5 | |
| GD1 alpha synthase, Sialyltransferase 7E, SIAT7 E, SIAT7E, ST6GalNAc V, ST6GALNAC5, ST6GalNAcV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26938, 365984, 81849 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BVH7, Q9QYJ1 | |
| St6galnac5 | |
| ST6GALNAC5 Fusion Protein Ag9767 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction