missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST6GALNAC5 Monoclonal Antibody (719508)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524336
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions. ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions (Tsuchida et al., 2003 ).
Specifications
| ST6GALNAC5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9BVH7 | |
| ST6GALNAC5 | |
| CHO-derived recombinant human SGalNAc alpha-2,6-sialyltransferase V/ST6GALNAC5 Gly30-Phe336. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 719508 | |
| Unconjugated | |
| ST6GALNAC5 | |
| AI851940; Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase V; alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase E; EC 2.4.99; EC 2.4.99.7; galNAc alpha-2,6-sialyltransferase V; GD1 alpha synthase; LOC100715749; MGC3184; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) E; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) E; sialyltransferase 7E; SIAT7E; SIAT7-E; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6 GalNAc alpha-2,6-sialyltransferase 5; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6GalNAc V; ST6GALNAC5; ST6GalNAcV | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 81849 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction