missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST6GALNAC2 Polyclonal Antibody
Description
Peptide sequence: VNTMKNSLVS YWNLGFTSVP QGQDLQYIFI PSDIRDYVML RSAILGVPVP Sequence homology: Cow: 91%; Dog: 85%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 100%; Rat: 100%.
ST6GALNAC2 belongs to a family of sialyltransferases that add sialic acids to the nonreducing ends of glycoconjugates. At the cell surface, these modifications have roles in cell-cell and cell-substrate interactions, bacterial adhesion, and protein targeting (Samyn-Petit et al., 2000 [PubMed 10742600]).
Specifications
Specifications
| Antigen | ST6GALNAC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ST6GALNAC2 |
| Gene Accession No. | Q9UJ37 |
| Gene Alias | (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase; (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II (); Gal-beta-1,3-GalNAc alpha-2,6-sialyltransferase; GalNAc alpha-2,6-sialyltransferase II; II; SAITL1; sialyltransferase 7; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-beta-galactosyl-1,3)-N-acetyl galactosaminde alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; sialyltransferase 7B; sialyltransferase-like 1; Siat7; SIAT7B; SIAT7-B; SIATL1; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 GalNAc alpha-2,6-sialyltransferase 2; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6GalNAc; ST6GalNAc II; St6galnac2; ST6GALNAC2, alpha-2,6-sialyltransferase 2; ST6GalNAcII; ST6GalNAII; SThM |
| Gene Symbols | ST6GALNAC2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?