missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST6GAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569666
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Transfers sialic acid from the donor of substrate CMP-sialic acid to galactose containing acceptor substrates. Has alpha-2,6-sialyltransferase activity toward oligosaccharides that have the Gal-beta-1,4-GlcNAc sequence at the non-reducing end of their carbohydrate groups, but it has weak or no activities toward glycoproteins and glycolipids.
Specifications
| ST6GAL2 | |
| Polyclonal | |
| Unconjugated | |
| ST6GAL2 | |
| alpha 2,6 sialyltransferase; alpha 2,6-ST 2; alpha-2,6-sialyltransferase; alpha-2,6-sialyltransferase ST6Gal II; beta galactoside alpha 2,6 sialyltransferase 2; beta-galactosamide alpha-2,6-sialyltransferase II; beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; C230064G14Rik; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; hST6Gal II; Kiaa1877; LOW QUALITY PROTEIN: beta-galactoside alpha-2,6-sialyltransferase 2; mKIAA1877; sialyltransferase; sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); SIAT2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2a; ST6 beta-galactoside alpha-2,6-sialyltransferase 2; ST6Gal II; st6gal2; st6gal2a; ST6GalII; ST6Gal-II | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 240119 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q76K27 | |
| ST6GAL2 | |
| synthetic peptide corresponding to a region of mouse ST6GAL2 (aa 286-335). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction