missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6 Sialyltransferase 6/ST6GALNAC6 Antibody (1F4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43040
This item is not returnable.
View return policy
Description
ST6 Sialyltransferase 6/ST6GALNAC6 Monoclonal antibody specifically detects ST6 Sialyltransferase 6/ST6GALNAC6 in Human samples. It is validated for Immunohistochemistry (Paraffin),ELISA,Immunohistochemistry,Western Blot
Specifications
| ST6 Sialyltransferase 6/ST6GALNAC6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 30815 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Western Blot | |
| 1F4 | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:500 | |
| alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6, CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)2,6-sialyltransferase member VI, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.7, GalNAc alpha-2,6-sialyltransferase VI, hST6GalNAc VI, RP11-203J24.3, Sialyltransferase 7F, sialytransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialytransferase) F, SIAT7F, SIAT7-F, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 6, ST6GalNAc VI, ST6GalNAcVI | |
| ST6(alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 - (Uniprot# Q969X2) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction