missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6 Sialyltransferase 2/ST6GALNAC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20494
This item is not returnable.
View return policy
Description
ST6 Sialyltransferase 2/ST6GALNAC2 Polyclonal specifically detects ST6 Sialyltransferase 2/ST6GALNAC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ST6 Sialyltransferase 2/ST6GALNAC2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B, alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2, EC 2.4.99.-, FLJ45660, GalNAc alpha-2,6-sialyltransferase II, SAITL1, sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B, sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase), sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase) B, Sialyltransferase 7B, sialyltransferase-like 1, SIAT7, SIAT7B, SIAT7-B, SIATL1, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 2, ST6GalNAc II, ST6GalNAcII, ST6GalNAII, SThM, STHM(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (89%), Dog (91%), Rat (87%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9UJ37 | |
| ST6GALNAC2 | |
| Recombinant protein encompassing a sequence within the center region of human ST6GALNAC2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10610 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction