missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6 Sialyltransferase 2/ST6GALNAC2 Antibody (025), Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
ST6 Sialyltransferase 2/ST6GALNAC2 Monoclonal antibody specifically detects ST6 Sialyltransferase 2/ST6GALNAC2 in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 025 |
| Dilution | ELISA |
| Regulatory Status | RUO |
| Target Species | Mouse |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?