missing translation for 'onlineSavingsMsg'
Learn More

ST6 Gal Sialyltransferase 2/ST6GAL2 Antibody [PerCP], Novus Biologicals™

Product Code. 30730868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30730868 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730868 Supplier Novus Biologicals Supplier No. AF7747C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ST6 Gal Sialyltransferase 2/ST6GAL2 Polyclonal antibody specifically detects ST6 Gal Sialyltransferase 2/ST6GAL2 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6 Gal Sialyltransferase 2/ST6GAL2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias Alpha 2,6-ST 2, beta-galactoside alpha-2,6-sialyltransferase 2, beta-galactoside alpha-2,6-sialyltransferase II, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2, EC 2.4.99, EC 2.4.99.1, FLJ30711, FLJ37730, FLJ38334, hST6Gal II, KIAA1877St6gal2, Sialyltransferase 2, sialyltransferase 2 (monosialoganglioside sialyltransferase), SIAT2, ST6 beta-galactosamide alpha-2,6-sialyltranferase 2, ST6Gal II, ST6GALII
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ST6 Gal Sialyltransferase 2/ST6GAL2, Glu243-Ser529, Accession # Q96JF0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84620
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.