missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Antibody Pair [HRP]

Product Code. 18609070 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18609070

Brand: Novus Biologicals™ NBP2796565Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.13 pg/mL
Assay Sensitivity 78.13-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N
Gene ID (Entrez) 6480
Gene Symbol ST6GAL1
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 83 ng of recombinant Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75.
  • Capture Antibody: 1 mg/mL of rabbit anti-Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Golgi Apparatus Markers
    Storage Requirements Storage is content dependent.
    Target ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.