missing translation for 'onlineSavingsMsg'
Learn More

ST3GAL6 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18409542 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409542 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409542 Supplier Novus Biologicals Supplier No. NBP177353V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ST3GAL6 Polyclonal antibody specifically detects ST3GAL6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL6
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias alpha2,3-sialyltransferase, CMP-NeuAc:beta-galactoside alpha-2,3-sialyltransferase VI, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.9, Sialyltransferase 10, SIAT10, ST3 beta-galactoside alpha-2,3-sialyltransferase 6, ST3Gal VI, ST3GalVI, ST3GALVIsialyltransferase 10 (alpha-2,3-sialyltransferase VI), type 2 lactosamine alpha-2,3-sialyltransferase
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ST3gal6. The immunogen is located within amino acids 230 - 280 of ST3gal6. Amino Acid Squence: PKVFPKNQKPKHPTTG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10402
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.