missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST3GAL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562056
This item is not returnable.
View return policy
Description
Immunogen sequence: SREDRYIELF YFPIPEKKEP CLQGEAESKA SKLFGNYSRD QPIFLRLEDY FWVKTPSAYE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
Synthesis of alpha-2, 3-linked sialic acid to Gal(beta-1, 3)GalNAc is mediated by at least 3 distinct beta-galactoside alpha-2, 3-sialyltransferases (EC 2.4.99.4), including ST3GAL4. In contrast, only a single gene encodes the beta-galactoside alpha-2, 6-sialyltransferase, ST6GAL1.
Specifications
| ST3GAL4 | |
| Polyclonal | |
| Unconjugated | |
| St3gal4 | |
| alpha 2,3-sialyltransferase IV; Alpha 2,3-ST 4; alpha-2,3-sialyltransferase; alpha2,3-sialyltransferase; alpha-2,3-sialyltransferase ST3Gal IV; alpha-3-N-acetylneuraminyltransferase; beta-galactoside alpha-2,3-sialyltransferase 4; CGS23; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4; FLJ11867; gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; glycosyltransferase; NANTA3; SAT3; SAT-3; sialyltransferase 4C; sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase); SIAT4; SIAT4C; SIAT4-C; ST3 beta-galactoside alpha-2,3-sialyltransferase 4; ST3Gal IV; ST3GAL4; ST3GalA.2; ST3GalIV; ST3GAL-IV; ST-4; STZ | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6484 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q11206 | |
| St3gal4 | |
| Recombinant protein corresponding to Human ST3GAL4. Recombinant protein control fragment (Product #RP-101187). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction