missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3GAL4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93783-0.1ml
This item is not returnable.
View return policy
Description
ST3GAL4 Polyclonal antibody specifically detects ST3GAL4 in Mouse samples. It is validated for Western Blot
Specifications
| ST3GAL4 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500 | |
| Alpha 2,3-sialyltransferase IV, Alpha 2,3-ST 4, Beta-galactoside alpha-2,3-sialyltransferase 4, CGS23FLJ46764, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.9, FLJ11867, gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, NANTA3alpha-3-N-acetylneuraminyltransferase, SAT3, SAT-3, Sialyltransferase 4C, sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase), SIAT4-C, SIAT4CCMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4, ST3 beta-galactoside alpha-2,3-sialyltransferase 4, ST3Gal IV, ST3GalA.2, ST3GalIV, ST-4, STZSIAT4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-267 of human ST3GAL4 (NP_006269.1). REDSFYFPIPEKKEPCLQGEAESKASKLFGNYSRDQPIFLRLEDYFWVKTPSAYELPYGTKGSEDLLLRVLAITSSSIPKNIQSLRCRRCVVVGNGHRLRNSSLGDAINKYDVVIRLNNAPVAGYEGDVGSKTTMRLFYPESAHFDPKVENNPDTLLVLVAFKAMDFHWIETILSDKKRVRKGFWKQPPLIWDVNPKQIRILNPFFMEIAADKLLSLPMQQPRKIKQKPTTGLLAITLALH | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 6484 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction