Learn More
Invitrogen™ ST3GAL3 Polyclonal Antibody

Description
Immunogen sequence: LHLLQWEEDS NSVVLSFDSA GQTLGSEYDR LGFLLNLDSK LPAELATKYA NFSEGACKPG YASALMTAIF PRFSKPAPMF LDDSFRKWAR IREFVPPFGI KGQDNLIKAI LSVTKEYRLT PALDSLRCRR CIIVGNGGVL ANKSLGSRID DYDIVVRLNS APVKGFEKDV GSKTTLRITY PEGAMQRPEQ YERDSLFVLA GFKWQDFKWL KYIVYKERVS ASDGFWKSVA TRVPKEPPEI RILNPYFIQE AAFTLIGLPF NNGLMGRGNI PTLGSVAVTM ALHGCDEVAV AGFGYDMSTP NAPLHYYETV RMAAIKESWT HNIQREKEFL RKLVKARVIT DLSSGI.
Specifications
Specifications
| Antigen | ST3GAL3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.52 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ST3GAL3 |
| Gene Accession No. | P97325, Q02734, Q11203 |
| Gene Alias | alpha 2,3-sialyltransferase III; alpha 2,3-ST 3; alpha-2,3-sialyltransferase II; beta-galactoside alpha-2,3-sialyltransferase 3; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase soluble form; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; LOW QUALITY PROTEIN: CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; EIEE15; gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase; Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase; mental retardation, non-syndromic, autosomal recessive, 12; MRT12; N-acetyllacosaminide alpha 2,3-sialyltransferase; N-acetyllactosaminide alpha-2,3-sialyltransferase; sialyltransferase (N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase 3; sialyltransferase 6; sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase); sialyltransferase 6(N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase ST3Gal-III; Siat3; Siat6; ST3 beta-galactoside alpha-2,3-sialyltransferase 3; ST3Gal III; St3gal3; ST3GALII; ST3GalIII; ST3N |
| Gene Symbols | ST3GAL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.