missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3GAL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-60067
This item is not returnable.
View return policy
Description
ST3GAL3 Polyclonal specifically detects ST3GAL3 in Human samples. It is validated for Western Blot.
Specifications
| ST3GAL3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ST3GAL3 | |
| Synthetic peptides corresponding to ST3GAL3(ST3 beta-galactoside alpha-2,3-sialyltransferase 3) The peptide sequence was selected from the C terminal of ST3GAL3. Peptide sequence GFGYDMSTPNAPLHYYETVRMAAIKESWTHNIQREKEFLRKLVKARVITD. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Pig: 92%; Zebrafish: 92%; Equine: 91%; Xenopus: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| alpha 2,3-sialyltransferase III, Alpha 2,3-ST 3, alpha-2,3-sialyltransferase II, Beta-galactoside alpha-2,3-sialyltransferase 3, CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase, EC 2.4.99.6, Gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase, Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase, N-acetyllactosaminide alpha-2,3-sialyltransferase, Sialyltransferase 6, SIAT6sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase), ST3 beta-galactoside alpha-2,3-sialyltransferase 3, ST3Gal III, ST3GALII, ST3GalIII, ST3N | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6487 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction