missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | ST3GAL3 |
| Synonym | alpha 2,3-sialyltransferase III, Alpha 2,3-ST 3, alpha-2,3-sialyltransferase II, Beta-galactoside alpha-2,3-sialyltransferase 3, CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase, EC 2.4.99.6, Gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase, Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase, N-acetyllactosaminide alpha-2,3-sialyltransferase, Sialyltransferase 6, SIAT6sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase), ST3 beta-galactoside alpha-2,3-sialyltransferase 3, ST3Gal III, ST3GALII, ST3GalIII, ST3N |
| Gene Symbol | ST3GAL3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?