missing translation for 'onlineSavingsMsg'
Learn More

ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Antibody, PerCP, Novus Biologicals™

Product Code. 18651482 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18651482 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18651482 Supplier Novus Biologicals Supplier No. NBP298478PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Polyclonal antibody specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.