missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£179.00 - £246.00
Specifications
| Antigen | ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1-2 μL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633462
|
Novus Biologicals
NBP2-98478-100ul |
100 μg |
£246.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680885
|
Novus Biologicals
NBP2-98478-50ul |
50 μg |
£179.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Polyclonal antibody specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Alpha 2,3-ST 2, Beta-galactoside alpha-2,3-sialyltransferase 2, beta-galactoside alpha-2,3-sialytransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2, EC 2.4.99.4, EC 2.4.99.7, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4B, sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase), SIAT4B, SIAT4-B, ST3 beta-galactoside alpha-2,3-sialyltransferase 2, ST3GalA.2ST3Gal II, ST3GALII | |
| Produced in rabbits immunized with E. coli-derived Human ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000, Immunoprecipitation 1-2 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 6483 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title