missing translation for 'onlineSavingsMsg'
Learn More

ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Antibody [Biotin], Novus Biologicals™

Product Code. 30732555 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732555 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732555 Supplier Novus Biologicals Supplier No. AF7275B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Polyclonal antibody specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Alpha 2,3-ST 2, Beta-galactoside alpha-2,3-sialyltransferase 2, beta-galactoside alpha-2,3-sialytransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2, EC 2.4.99.4, EC 2.4.99.7, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4B, sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase), SIAT4B, SIAT4-B, ST3 beta-galactoside alpha-2,3-sialyltransferase 2, ST3GalA.2ST3Gal II, ST3GALII
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ST3 β-Gal alpha -2,3-Sialyltransferase 2/ST3GAL2, Pro52-Gln350, Accession # NP_008858
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6483
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.