missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31449
This item is not returnable.
View return policy
Description
ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A Polyclonal specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7), 1% BSA, 20% Glycerol with 0.025% Proclin 300 | |
| ST3GAL1 | |
| Recombinant protein encompassing a sequence within the center region of human SIAT4A. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 6482 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.98mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Alpha 2,3-ST 1, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1, DKFZp666E036, DKFZp779K2051, EC 2.4.99, EC 2.4.99.4, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4A, sialyltransferase 4A (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4A (beta-galactoside alpha-2,3-sialyltransferase), sialyltransferase 4A (beta-galactoside alpha-2,3-sialytransferase), SIAT4, SIAT4A, SIAT4-A, SIATFLFLJ36548, ST3 beta-galactoside alpha-2,3-sialyltransferase 1, ST3Gal I, ST3GalA.1MGC9183, ST3GalI, ST3GalIA, ST3GalIA1,Beta-galactoside alpha-2,3-sialyltransferase 1, ST3OST3GalA | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction