missing translation for 'onlineSavingsMsg'
Learn More

ST271, MedChemExpress

Product Code. 16496517
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496517 5 mg 5mg
16494846 100 mg 100mg
16443798 10 mg 10mg
16440177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496517 Supplier Medchem Express Supplier No. HY103097
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ST271 is a potent inhibitor of protein tyrosine kinase (PTK), inhibits phospholipase D activation stimulated by fMet-Leu-Phe and PAF, with IC50s of 6.7 and 9 μM, respectively.

ST271 is a potent inhibitor of protein tyrosine kinase (PTK), inhibits phospholipase D activation stimulated by fMet-Leu-Phe and PAF, with IC50s of 6.7 and 9 μM, respectively.

Specifications

Chemical Name or Material ST271
CAS 106392-48-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H20N2O2
Quantity 5 mg
Solubility Information DMSO : ≥ 110 mg/mL (403.91 mM)
SMILES O=C(N)/C(C#N)=C/C1=CC(C(C)C)=C(O)C(C(C)C)=C1
Molecular Weight (g/mol) 272.34
Formula Weight 272.34
Percent Purity 98.9%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.