missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11920-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes the 80 kDa, 40 kDa, and 30 kDa forms in different tissues and cells.
ST2 is a member of the interleukin 1 receptor family. Studies of ST2 in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. ST2 belongs to the interleukin-1 receptor family with three Ig-like C2-type (immunoglobulin-like) domains and a TIR domain. ST2 is a receptor for interleukin-33 (IL-33) which upon stimulation recruits the signaling molecules MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. Reports also suggest its possible role in helper T-cell function as well as in cell growth. Three isoforms of ST2 have been reported where isoform C is a membrane form and isoform B is a secreted form. ST2 may be induced by proinflammatory stimuli, and may also be found in serum of autoimmune disease. Different isoforms are restricted to different tissues while high expression of this protein is evident in kidney, lung, placenta, stomach, skeletal muscle, colon and small intestine.Specifications
| ST2 | |
| Polyclonal | |
| Unconjugated | |
| IL1RL1 | |
| DER4, FIT 1, IL1RL1, interleukin 1 receptor like 1, Protein ST2, ST2, ST2L, ST2V, T1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17082, 25556, 9173 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14719, Q01638, Q62611 | |
| Il1rl1 | |
| ST2 Fusion Protein Ag2555 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction