missing translation for 'onlineSavingsMsg'
Learn More

ST-193, MedChemExpress

Product Code. 16496087
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496087 5 mg 5mg
16484606 100 mg 100mg
16460759 1 mg 1mg
16453368 10 mg 10mg
16449896 50 mg 50mg
16422736 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16496087 Supplier Medchem Express Supplier No. HY101441
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ST-193 is a potent broad-spectrum arenavirus inhibitor; inhibits Guanarito, Junin, Lassa and Machupo virus with IC50 values of 0.44, 0.62, 1.4 and 3.1 nM, respectively.

ST-193 is a potent broad-spectrum arenavirus inhibitor

Specifications

Chemical Name or Material ST-193
CAS 489416-12-8
Color Pink
Purity Grade Notes Research
Molecular Formula C24H25N3O
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (269.20 mM; Need ultrasonic)
SMILES CC(C1=CC=C(CNC2=CC=C3C(N=CN3C4=CC=C(OC)C=C4)=C2)C=C1)C
Molecular Weight (g/mol) 371.47
Formula Weight 371.47
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.