missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSX6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568722
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
This gene belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This gene is classified as a pseudogene because a splice donor in the 3' UTR has changed compared to other family members, rendering the transcript a candidate for nonsense-mediated mRNA decay (NMD).
Specifications
| SSX6 | |
| Polyclonal | |
| Unconjugated | |
| SSX6 | |
| dJ54B20.1; psiSSX2; SSX family member 6, pseudogene; SSX6; SSX6P; SSXP2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 280657 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7RTT6 | |
| SSX6 | |
| synthetic peptide directed towards the middle region of human SSX6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction