missing translation for 'onlineSavingsMsg'
Learn More

SSX4 Antibody (3E10), Novus Biologicals™

Product Code. 18368259 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368259 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368259 Supplier Novus Biologicals Supplier No. H00006759M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SSX4 Monoclonal antibody specifically detects SSX4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSX4
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3E10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005627
Gene Alias Cancer/testis antigen 5.4, CT5.4SSX4B, MGC119056, MGC12411, protein SSX4, SSX4A, synovial sarcoma, X breakpoint 4
Host Species Mouse
Immunogen SSX4 (NP_005627, 91 a.a. - 188 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RNQVERPQMTFGSLQRIFPKIMPKKPAEEENGLKEVPEASGPQNDGKQLCPPGNPSTLEKINKTSGPKRGKHAWTHRLRERKQLVVYEEISDPEEDDE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6759
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.