Learn More
SSX2 Antibody (CL3202) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-42398AF750
Product Code. 30305065
  • £558.00 / 0.10mL
Description
SSX2 Monoclonal antibody specifically detects SSX2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SSX2
Monoclonal
Alexa Fluor 750
Cancer/testis antigen 5.2, cancer/testis antigen family 5, member 2a, CT5.2a, HD21, HOM-MEL-40, MGC119055, MGC15364, MGC3884, protein SSX2, sarcoma, synovial, X-chromosome-related 2, SSX, SSX2A, SSX2B, synovial sarcoma, X breakpoint 2B, synovial sarcoma, X breakpoint 2CT5.2, Tumor antigen HOM-MEL-40
This antibody was developed using a recombinant protein derived from Q16385, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
6757
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3202
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified