missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551082
This item is not returnable.
View return policy
Description
SSX1 Polyclonal Antibody detects endogenous levels of SSX1 protein.
Could act as a modulator of transcription.
Specifications
| SSX1 | |
| Polyclonal | |
| Unconjugated | |
| SSX1 | |
| Cancer/testis antigen 5.1; cancer/testis antigen family 5, member 1; CT5.1; CT5.2; CT51; Protein SSX1; sarcoma, synovial, X-chromosome-related 1; SSRC; SSX family member 1; SSX1; SSX2; SSX2A; SSX2B; SSX3; Synovial sarcoma, X breakpoint 1; X breakpoint 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6756 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| Q16384 | |
| SSX1 | |
| Synthesized peptide derived from the C-terminal region of human SSX. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction