missing translation for 'onlineSavingsMsg'
Learn More

SSX1 Antibody (OTI1E10), DyLight 594, Novus Biologicals™

Product Code. 18407647 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407647 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407647 Supplier Novus Biologicals Supplier No. NBP274362DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SSX1 Monoclonal antibody specifically detects SSX1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSX1
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1E10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Cancer/testis antigen 5.1, CT5.1cancer/testis antigen family 5, member 1, MGC150425, protein SSX1, sarcoma, synovial, X-chromosome-related 1, SSRC, synovial sarcoma, X breakpoint 1MGC5162
Host Species Mouse
Immunogen Full length recombinant protein of human SSX1 (NP_005626) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6756
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.