missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSUH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562042
This item is not returnable.
View return policy
Description
Immunogen sequence: AKAKGENLFK DENSVVYPIV DFPLRDISLA SQRGIAEHSA ALASRARVLQ QRQTIELIPL TEVHYWYQGK TYVYYIYGTD HQVYAVDYPE RYCCGCT Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 42%.
SSUH2 is a protein coding gene.
Specifications
| SSUH2 | |
| Polyclonal | |
| Unconjugated | |
| SSUH2 | |
| C20H3orf32; C22H3orf32; C3orf32; D630042P16Rik; Fls485; Protein ssu-2 homolog; protein SSUH2 homolog; RGD1562010; Ssu2; SSU-2; ssu-2 homolog; ssu-2 homolog (C. elegans); SSUH2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51066 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2M2 | |
| SSUH2 | |
| Recombinant protein corresponding to Human SSUH2. Recombinant protein control fragment (Product #RP-100135). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction