missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSTR5 Recombinant Rabbit Monoclonal Antibody (24GB2600)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557729
This item is not returnable.
View return policy
Description
SSTR5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predominantly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.
Specifications
| SSTR5 | |
| Recombinant Monoclonal | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08858, P30938, P35346 | |
| Sstr5 | |
| A synthesized peptide derived from human SSTR5 (aa 1-64). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2600 | |
| Unconjugated | |
| Sstr5 | |
| Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20609, 25354, 6755 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction