missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSTR4 Recombinant Rabbit Monoclonal Antibody (HL2933)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556881
This item is not returnable.
View return policy
Description
SSTR4 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
SSTR4 has been reported to be expressed in brain, gastrointestinal tract, pancreas, and prostate tissues. No ESTs have been identified.
Specifications
| SSTR4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P31391 | |
| SSTR4 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human SSTR4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2933 | |
| Unconjugated | |
| SSTR4 | |
| G-protein coupled receptor; Smstr4; somatostatin receptor; somatostatin receptor 4; Somatostatin receptor subtype 4 Rattus norvegicus Sprague-Dawley major hippocampal somatostatin receptor (SSTR4) mRNA; somatostatin receptor type 4; SS4R; SS-4-R; SS4-R; sst4; Sstr4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6754 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction