missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSTR4 Recombinant Rabbit Monoclonal Antibody (7H49L61)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703478
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Mouse, Rat.
SSTR4 has been reported to be expressed in brain, gastrointestinal tract, pancreas, and prostate tissues. No ESTs have been identified.
Specifications
| SSTR4 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P31391 | |
| SSTR4 | |
| Peptides corresponding to human SSTR4 [1) aa367-aa388 2) aa2-aa20]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Immunocytochemistry | |
| 7H49L61 | |
| Unconjugated | |
| SSTR4 | |
| G-protein coupled receptor; Smstr4; somatostatin receptor; somatostatin receptor 4; Somatostatin receptor subtype 4 Rattus norvegicus Sprague-Dawley major hippocampal somatostatin receptor (SSTR4) mRNA; somatostatin receptor type 4; SS4R; SS-4-R; SS4-R; sst4; Sstr4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6754 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction