Learn More
Invitrogen™ SSTR2 Recombinant Rabbit Monoclonal Antibody (HL3078)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556879
Product Code. 30275974
  • £434.00 / 100µL
Description
SSTR2 Recombinant Monoclonal Antibody for ICC/IF, IHC (P)
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SSTR2
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P30874
SSTR2
Synthetic peptide encompassing a sequence within the Extracellular domain of human Somatostatin receptor 2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Immunocytochemistry
HL3078
Unconjugated
SSTR2
Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A
Rabbit
Protein A
RUO
6752
-20°C, Avoid Freeze/Thaw Cycles