missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSTR2 Recombinant Human Monoclonal Antibody (7A9)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550074
This item is not returnable.
View return policy
Description
SSTR2 Recombinant Monoclonal Antibody for Flow, ELISA
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
Specifications
| SSTR2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P30874 | |
| SSTR2 | |
| Recombinant Human SSTR2 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Neutralization | |
| 7A9 | |
| Unconjugated | |
| SSTR2 | |
| Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 6752 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction