Learn More
SSTR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20404-1-AP-20UL
 View more versions of this product
Product Code. 16899934
  • £135.00 / 20µL
Description
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
Specifications
Specifications
SSTR2
Polyclonal
Unconjugated
SSTR2
somatostatin receptor 2, Somatostatin receptor type 2, SRIF 1, SS 2 R, SS2 R, SS2R, SSTR2
Rabbit
Antigen Affinity Chromatography
RUO
6752
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P30874
SSTR2
SSTR2 Fusion Protein Ag14205
20 μL